Would you like to log in again?
F.C. Bond Ball Mill, Bico
Supplier: Bico Braun International
REVIEWS (0)
Total Ratings: 0
Avg. Ratings: out of 5
Sort all by:
395-50
20058-110EA 26726.97 USD InStock
20058-110
F.C. Bond Ball Mill, Bico
Mills
Designed by F.C. Bond for use by cement companies and quality control laboratories throughout the world. Can be used in calculating the grindability of all ore.
Ordering information: 285 Ball Charge. Contact your Avantor sales representative for more information.
Order Now
ORDER
WRITE A REVIEW
REVIEWS (0)
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty