Cadence Science® hypodermic needles are precision made and designed to ensure strict adherence with U.S
Nonsterile. Packaged 12 needles per box.
Government specification GG-N-196. Points are machine ground to achieve uniform sharpness. Precision grinding, micro-bead jet blasting, and ultrasonic cleaning assure freedom from burrs and debris. All needle cannulae are fabricated from type 304 stainless steel tubing which must pass our rigorous in-plant quality control prior to acceptance and use.
Certifications: Plated brass luer hubs comply with luer taper specifications ISO 594.