Would you like to log in again?
Due to scheduled system maintenance, vwr.com will be temporarily unavailable from 6:00 am EST to 2:00 PM EST on Saturday, May 25. 
Reusable Animal Feeding/Oral Gavage Needles, Cadence Science®
Supplier: Cadence Science
7911 7908 7900 7906 7916 7910 7914 7922 7909 7920 7902 7917 7913 7924 7905 7921 7915 7923 7901 7912 7904 7903 7946 7919
89233-526PK 923.32 USD InStock
89233-526 89233-528 20068-640 20068-650 20068-642 20068-620 20068-630 20068-652 89233-530 20068-644 20068-622 20068-610 20068-624 20068-646 20068-612 20068-648 20068-636 20068-614 20068-606 20068-628 20068-638 20068-616 20068-619 20068-608
Reusable Animal Feeding/Oral Gavage Needles, Cadence Science®
Animal Supplies Animal Feeding Needles
Alll-metal, non-flexible stainless steel reusable gavage needles are designed for oral gavage procedures in laboratory animals.
  • Cadence Science® reusable feeding needles are available in two basic styles - straight and curved
  • Variety of gauges and lengths is available for different sized animals
  • Can be cleaned and re-used as needed
  • Non-sterile, packaged and sold in dozen quantity
The precision assembly of stainless steel balls and 304 stainless steel tubing ensures a quality needle suited for feeding and dosing without causing trauma to the mouth or esophagus. Ideal for pharmacologists, toxicologists, pathologists and other biomedical researchers working with laboratory animals.
Ordering information: Needles are supplied nonsterile and packaged 12 needles per box.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty