Would you like to log in again?
KIMAX® Specific Gravity Bottle, Le Chatelier, Kimble Chase
Supplier: Kimble Chase
KIMAX®
15115-24
20110-001CS 437.05 USD InStock
20110-001
KIMAX® Specific Gravity Bottle, Le Chatelier, Kimble Chase
Specific Gravity Bottles
KIMAX® bottle is used to determine the specific gravity of cement, sand, and other fine materials (ASTM C188)
Designed for use with a 64g sample. Body holds approximately 250mL, and the smaller neck bulb holds 17mL. The neck is graduated from 0–1.0mL and has two extra 0.1mL lines at both 1.0mL and 0.0mL. Graduated from 18–24mL in 0.1mL subdivisions. All markings are durable white enamel. Supplied with serial number. Designed from ASTM Specification E694 and complies with C188.
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty