Would you like to log in again?
VWR® Disposable Microcentrifuge Tubes
Supplier: VWR
VWR®
CFT458005 CFT457020 CFT451015 CFT450015 CFT454015 CFT458020 CFT458015 CFT455020 CFT450020 CFT454005 CFT456005 CFT451020 CFT452015 CFT455015 CFT454020 CFT452020 CFT453005 CFT450005 CFT453020 CFT455005 CFT456020 CFT451005 CFT452005 CFT457005 CFT456015 CFT457015 CFT453015
89000-040CS 330.42 USD InStock
89000-040 89000-028 20170-084 20170-088 20170-086 89000-020 89000-042 89000-044 89000-022 89000-024 89000-026 89000-030 20170-094 89000-018 20170-170 20170-092 20170-098 20170-096 89000-032 89000-010 89000-012 89000-034 89000-036 20170-090 89000-014 89000-038 89000-016
VWR® Disposable Microcentrifuge Tubes
Tubes Microcentrifuge Tubes
These tubes are made from USP Class VI PP.
  • Amber colored for light-sensitive substances
  • RNase-, DNase-, and endotoxin-free
  • Engraved graduations ensure accuracy
  • Caps can be penetrated by a syringe and will not pop open during centrifuging
These tubes also feature flat, pierceable, frosted caps and a side labeling surface.
Autoclavable at 121 °C and freezable to –80 °C.
Certifications: Manufactured in a Class 100,000 cleanroom environment. Manufactured under ISO13485:2016 and ISO9001:2015 quality management systems. Meet the requirements of RoHS.
Ordering information: Assorted pack contains 100 tubes each of red, orange, yellow, green and blue.
Order Now
ORDER
SPECIFICATIONS
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty