Would you like to log in again?
Microcentrifuge Tubes
Supplier: Bio Plas
REVIEWS (0)
Total Ratings: 0
Avg. Ratings: out of 5
Sort all by:
4204 4222R 4217R 4202G 4221R 4223R 4203 4203 4219R 4215R 4200 4220R 4201 4224R 4222R 4202 4217R 4216R 4202 4221R 4223R 4220R 4218R 4224R 4204 4215R 4216R 4218R 4201
20170-705PK 87.27 USD InStock
20170-705 20170-704 20170-702 20170-709 20170-708 20170-789 20170-723 20170-700 20170-788 20170-787 20170-786 77148-114 20170-781 20170-780 20170-713 20170-774 20170-773 20170-772 20170-793 20170-771 20170-778 20170-777 20170-710 20170-776 20170-775 20170-770 20170-792 20170-791 20170-790
Microcentrifuge Tubes
Tubes Microcentrifuge Tubes
Ideal for applications in freezer storage, autoclaving, boiling, or irradiation.
  • External ridges fit rack serrations and allow one-handed opening
Colored PPCO screw caps are fitted with an ethylene propylene O-ring for reliable, leakproof performance. Compatible with most microcentrifuges with fixed angle heads. Withstands over 20000×g.
Tubes are made of reusable polypropylene copolymer (PPCO). Seamless conical construction with squared external threads prevents hang up of samples and reduces contamination.
Ordering information: Dual-ended polypropylene tool 20170-688 provides a combination of socket wrench and bottle top action for quick, effective tube access. Tubes and caps are available separately.
Order Now
ORDER
ACCESSORIES
WRITE A REVIEW
REVIEWS (0)
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty