Would you like to log in again?
IOWA STATE CONDENSER
Supplier: HGF GLASS, INC
Condenser^ 200mm, 28/15 socket joint for Iowa state distilling apparatus which is used for determinations in the citrus indusctry, but applicable to other distilling procedures.
ORDER
Expand All / Collapse All
Description VWR Catalog Number Supplier No Unit Price Quantity
IOWA STATE CONDENSER 20203-006 210200-2 Each Retrieving
Generate Barcodes Barcode Label Format: 
Generate PDF Catalog Page
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty