Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Flex-Tubes® Microtubes, 1.5 ml

Supplier: Eppendorf
Flex-Tubes®
022363557 022363590 022363590 022363573 022363514 022364120 022363573 022364111 022364120 022363514 022363531 022363557 022364111 022363531
62111-756PK 81.62 USD InStock
62111-756 CA20901-427 20901-427 CA62111-756 CA20901-430 CA20901-551 20901-430 20901-551 20901-421 CA20901-434 20901-424 20901-434 CA20901-424 CA20901-421
Flex-Tubes® Microtubes, 1.5 ml
Tubes Microcentrifuge Tubes
Flex-Tube® microtubes (1.5 ml) are designed for seamless one-handed operation with an easy-to-open, securely closing tube design. Perfect for sample preparation, centrifugation, and storage, these tubes will effortlessly integrate into workflows for efficient and reliable performance.

  • Easy-to-use ergonomic design featuring a user-friendly tube lid
  • Durable polypropylene construction capable of withstanding chemicals, mechanical stress, and extreme temperatures
  • g-Safe® technology ensures stability up to 30000×g, preventing sample loss during centrifugation and enhancing safety with hazardous samples
  • Free from slip agents, plasticizers, and biocides; eliminating substances that may compromise bio-assay results
  • Clear graduation marks allow for visual volume verification during pipetting
  • Convenient frosted lid and side surface facilitate easy labeling
  • Functional across a wide temperature range (−86 to 100 °C) for versatile applications
  • Autoclavable up to 121 °C for 20 minutes, ensuring effective sterilization
  • Available in both Eppendorf Quality and PCR Clean purity grades with lot certification

The easy-to-open, securely closing, Flex-Tube® offers comfort and dependability for sample preparation, centrifugation, and storage. One vessel for all applications in molecular biology, chemistry, and biochemistry.

Eppendorf consumables, including the Flex-Tube®, are made exclusively from high-quality, ultra-pure polypropylene, which complies with FDA guidelines 21 CFR § 177.1520 »Olefin Polymers«, and 21 CFR § 178.2010 »Antioxidants and Stabilizers for Polymers«. Extensive quality checks are in place prior to and during production. Furthermore, the final product is examined for functionality.

The functionality tests for tubes include lid closure security in a boiling water bath, centrifugation stability, and vapor tightness. These quality controls and tests ensure the sustained high product quality across the entire production phase - lot by lot.

Highest sample integrity.
Manufactured without the use of slip agents, plasticizers, and biocides - substances that have been shown to leach from plastic consumables into the sample and negatively affect bio-assay results or measurements, e.g. nucleic acid quantification by photometric measurements at 260 nm.

Certifications: Certificate of Quality/Conformity (ISO 9001, ISO 13485, ISO 14001), Lot-specific certficates for the purity grade PCR clean can be downloaded from the Eppendorf website.

Order Now
 

ORDER

SPECIFICATIONS

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty