Would you like to log in again?
Eppendorf® IsoTherm Benchtop Cooler
Supplier: Eppendorf
022510291 022510169 022510185 022510282 022510100 022510282 022510142 022510002
20901-648PK 231.19 USD InStock
20901-648 20901-659 CA20901-659 20901-647 20901-651 20901-661 20901-650 20901-645
Eppendorf® IsoTherm Benchtop Cooler
Coolers Benchtop Coolers
IsoTherm cooler eliminates the messiness and contamination risk of ice buckets while protecting temperature-sensitive samples and enzymes during use on the benchtop
Starter sets are a complete system consisting of one 0°C (32°F) IsoPack cold pack, one –21°C (–5.8°F) IsoPack cold pack, one transparent IsoRack, and one IsoSafe insulated transport rack. IsoPack and IsoRack sets may also be purchased separately.
The system can maintain 0°C (32°F) up to 6 hours and –21°C (–5.8°F) for 3 hours. It utilizes a modular rack system numbered for easy sample storage and identification. Each autoclavable IsoRack holds twenty-four 0.5mL or 1.5–2.0mL tubes, withstands temperatures down to –200°C (–328°F), floats in a water bath while thawing or incubating samples, and can be centrifuged in a microplate rotor.
Order Now
ORDER
ACCESSORIES
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty