Would you like to log in again?
SP Bel-Art Microcentrifuge Tube Rack, Bel-Art Products, a part of SP
Supplier: Bel-Art
REVIEWS (0)
Total Ratings: 0
Avg. Ratings: out of 5
Sort all by:
SCIENCEWARE®
F18843-0000
20901-895CS 208.15 USD InStock
20901-895
SP Bel-Art Microcentrifuge Tube Rack, Bel-Art Products, a part of SP
Racks Tube Racks
This two-tier polypropylene rack holds twenty 1.5 mL microcentrifuge tubes in numbered wells.
  • Polypropylene construction
  • Easy handling with suspended holding design
  • Autoclavable
Tubes are held with roughly 25 mm (1") protruding above the rack surface for easy handling. The rack is steam autoclavable at 121°C (250°F).
Order Now
ORDER
WRITE A REVIEW
REVIEWS (0)
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty