Would you like to log in again?
SP Bel-Art Microcentrifuge Floating Rack, Bel-Art Products, a part of SP
Supplier: Bel-Art
REVIEWS (0)
Total Ratings: 0
Avg. Ratings: out of 5
Sort all by:
F18876-0000
20901-905CS 366.04 USD InStock
20901-905
SP Bel-Art Microcentrifuge Floating Rack, Bel-Art Products, a part of SP
Racks Tube Racks
Incubate up to 24 microcentrifuge tubes in a water bath (choose from 1.5 mL or 0.5 mL size).
  • Floating rack design
  • Easy to load
  • Snug fit for secure holding of microtubes
Easy to load, the microtubes are held snugly and then released when the rack is pressed down on a benchtop.
Order Now
ORDER
DOCUMENTATION
WRITE A REVIEW
REVIEWS (0)
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty