Would you like to log in again?
Upchurch Scientific® Fittings Kits, IDEX Health and Science
Supplier: IDEX HEALTH & SCIENCE LLC
Upchurch Scientific®
1454 1420 1421 1457 1453 1412 1413 1356 1451 1456 1404 1452
21511-372EA 577.67 USD InStock
21511-372 21511-370 53500-666 53498-566 21001-112 53498-532 53500-848 21001-110 21511-368 21001-116 21001-114 53500-610
Upchurch Scientific® Fittings Kits, IDEX Health and Science
Fittings and Connectors
MicroTight™ Tubing Sleeves facilitate capillary tubing connections with our MicroTight accessories
Manufacturer Specific Fittings Kits are customized for specific manufacturer’s legacy HPLC systems.
IDEX Health & Science manufactures a large selection of kits, each containing an assortment of products designed to make your life easier and to save you money. Our kits range from our Super Kit – which contains a collection of many products you’ll use to accessorize your systems – to more specialized kits like our FIA kit, offering products designed to help you with flow injection analyses.
Sleeves feature an outer diameter of .025" and are available in variety of inner diameters for a broad-range compatibility with a wide assortment of capillary tubing sizes. Manufactured from PEEK polymer, these sleeves carry an upper temperature threshold of 125 °C. For proper sleeve use, choose a sleeve with an inner diameter .001"-.002" (25-50 µm) larger than the outer diameter of your capillary tubing. Then slip the sleeve over the flow path tubing, such that the tubing extends all the way through the sleeve, but not beyond the end of the sleeve. Choose the correct fitting that corresponds with your receiving port, slide it over the sleeved flow path tubing and connect as normal.
Certifications: ROHS Certified
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty