Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Deluxe Pallet Trucks, Wesco

Supplier: Wesco

REVIEWS (0)

Total Ratings: 0
Avg. Ratings: out of 5

Sort all by:

272748 272765
21002-052EA 1828.25 USD InStock
21002-052 21002-054
Deluxe Pallet Trucks, Wesco
Carts
Designed for economical, convenient lifting, transporting, and lowering of wooden pallets.

  • Pump overload bypass
  • Rubber-coated safety loop handle features a triple function hand control (raise, neutral, lower) to maneuver easily in any position
  • Adjustable fork tie rods
  • Tapered fork tips with rugged entry guides help reduce damage
  • Box-formed, 8-gauge, 15.2 cm (6") wide, high-tensile steel forks are structurally reinforced at critical stress points to help prevent bending
  • Large mold-on polyurethane steering wheel allows smooth rolling over uneven surfaces
  • A 200° steering angle allows extra maneuverability in tight areas
  • Exit rollers

Fork footprint: 27x48"

Ordering information: Manufacturer's five-year warranty on pump.

Order Now
 

ORDER

ACCESSORIES

WRITE A REVIEW

REVIEWS (0)

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty