Would you like to log in again?
Scheduled maintenance of the site is planned from June 8th at 7:00 PM to June 9th at 5:00 PM ET. During this time, contract pricing and registration functionality will be unavailable. We appreciate your patience.
DRUM PLUG WRENCH UNIVERSAL
Supplier: WESCO INDUSTRIAL PRODUCTS, LLC
Universal Drum Plug Wrench^ Heavy-duty cast iron construction. Opens most plugs and bung nuts, male and female, and steel and plastic. Iron alloy will not chip or bend in normal use. Handles bung patterns 1, 2, 3, and 4.
ORDER
Expand All / Collapse All
Description VWR Catalog Number Supplier No Unit Price Quantity
DRUM PLUG WRENCH UNIVERSAL 21002-056 272017 Each Retrieving
Generate Barcodes Barcode Label Format: 
Generate PDF Catalog Page
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty