Would you like to log in again?
Labcor™ Pipettor Solution Basins, Biotix
Supplier: Biotix
Labcor™
730-001 730-014 730-016 730-004 730-006 730-011
89092-832CS 126.85 USD InStock
89092-832 89092-834 89092-836 89092-838 21007-972 21007-970
Labcor™ Pipettor Solution Basins, Biotix
Reagent Reservoirs
Disposable basins are designed for 8- and 12-channel pipettors
  • Angled sides and V-shaped bottom permit easy access to solution
  • Pre-sterile basins are electron beam sterilized and RNase-, DNase-, and pyrogen-free
  • Polyvinyl chloride material allows the contents to be clearly viewed
Basins are available pre-sterile or nonsterile and constructed of either polyvinyl chloride or polystyrene.
Order Now
ORDER
ACCESSORIES
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty