Would you like to log in again?
Corning® Microcentrifuge Tubes with Screw Caps, Polypropylene, Sterile, Corning
Supplier: Corning
REVIEWS (0)
Total Ratings: 0
Avg. Ratings: out of 5
Sort all by:
Corning®
430917 430915 430909
21008-802CS 336.23 USD InStock
21008-802 21008-809 21008-807
Corning® Microcentrifuge Tubes with Screw Caps, Polypropylene, Sterile, Corning
Tubes Microcentrifuge Tubes
Polypropylene, with orange screw cap.
  • Good chemical resistance
  • Good temperature resistance
  • Screw caps with O-rings provide a tight secure seal
  • With large white marking spot
Order Now
ORDER
DOCUMENTATION
WRITE A REVIEW
REVIEWS (0)
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty