Would you like to log in again?
Please be aware that our e-commerce platform will be undergoing scheduled maintenance from 6:00 AM to 2:00 PM ET on May 11, 2024. During this time our site will be not available. We appreciate your patience.
Nalgene® Oak Ridge Centrifuge Tubes, Polycarbonate, Thermo Scientific
Supplier: Thermo Scientific
Nalgene®
3138-0030 3138-0016 3118-0085 3118-0028 3138-0050 3118-0010 3118-0030 3118-0050
21010-385CS 1223.68 USD InStock
21010-385 21010-343 21009-310 21009-353 21009-331 21009-320 21009-342 21010-400
Nalgene® Oak Ridge Centrifuge Tubes, Polycarbonate, Thermo Scientific
Tubes Centrifuge Tubes
Transparent autoclavable polycarbonate tubes.
  • Polypropylene screw caps
  • Leakproof closures provide safe containment
  • Withstand up to 50000 ×g in standard and refrigerated centrifuges
Place cap loosely on tube when autoclaving, without engaging threads.
Note: For proper performance, fill tubes to at least 80% of the total capacity.
Certifications: Meet OSHA Standard 29 CFR Part 1910.1030 for use as protection against bloodborne pathogens.
Ordering information: Tubes with sealing caps include a silicone gasket in addition to the tube and screw closure. Nominal volumes correspond to rotor tube sizes; please refer to nominal volumes when ordering tubes to fit a rotor.
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty