Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Nalgene® Oak Ridge Centrifuge Tubes, Teflon® FEP, Thermo Scientific

Supplier: Thermo Scientific
Nalgene®
3114-0010 3114-0050 3114-0030
21009-422CS 576.57 USD InStock
21009-422 21009-477 21009-455
Nalgene® Oak Ridge Centrifuge Tubes, Teflon® FEP, Thermo Scientific
Tubes Centrifuge Tubes
For high-speed centrifugation of aggressive chemicals.

  • Autoclavable and leakproof
  • Resists any acid, base, or solvent
  • Withstand temperatures from –100 to 150 °C (–148 to 302 °F)
  • Withstand upto 50000 ×g in refrigerated centrifuges

Excellent for lipid and phenol extractions, as well as biomolecular applications using nucleic acids, proteins, and more. Provide storage for expensive or aggressive samples. Before autoclaving, place closure on container without engaging threads.

Note: FEP centrifuge ware must be completely filled and refrigerated for centrifugation. Not to be used at room temperature.

Certifications: Meet OSHA Standard 29 Part 1910.1030 for use as protection against bloodborne pathogens.

Ordering information: Supplied with Tefzel® ETFE screw closures. NALGENE® sealing cap assemblies may be ordered separately. Nominal volumes correspond to rotor tube sizes; please refer to nominal volumes when ordering tubes to fit a rotor.

Order Now
 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty