Would you like to log in again?
Nalgene® Sealing Caps for Oak Ridge Centrifuge Tubes, Thermo Scientific
Supplier: Thermo Scientific
DS3132-0020 DS3132-0024
21010-003CS 41.39 USD InStock
21010-003 21010-025
Nalgene® Sealing Caps for Oak Ridge Centrifuge Tubes, Thermo Scientific
Closures Bottle Closures
Oak Ridge style sealing caps ensure leak proof service of Nalgene centrifuge tubes and bottles at their recommended maximum speeds (10,000 x g or greater), even when full
Also recommended for spinning hazardous materials compatible with centrifuge tube or bottle material. Before autoclaving, set closure on top of the container without engaging the threads. Fits Oak ridge Tube Size is 28 mL and 30 mL for 21010-003 series, 50 mL for 21010-025 series.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty