Would you like to log in again?
Please be aware that our e-commerce platform will be undergoing scheduled maintenance from 6:00 AM to 2:00 PM ET on May 11, 2024. During this time our site will be not available. We appreciate your patience.
Nalgene® Oak Ridge Centrifuge Tubes, Polysulfone, Thermo Scientific
Supplier: Thermo Scientific
Nalgene®
3115-0050 3115-0030 3137-0050
21010-501CS 1545.23 USD InStock
21010-501 21009-660 21009-681
Nalgene® Oak Ridge Centrifuge Tubes, Polysulfone, Thermo Scientific
Tubes Centrifuge Tubes
Strong plastic tubes with good resistance to corrosive bases.
  • Leakproof and autoclavable
  • Transparent with a yellow tint
  • Maximum RCF is 50000 ×g
  • Polypropylene screw closures
For standard and refrigerated centrifugation of biological and chemical samples. When autoclaving, place cap loosely on tube without engaging threads.
Note: For proper performance, fill tubes to at least 80% of the total capacity.
Certifications: Meet OSHA Standard 29 CFR Part 1910.1030 for use as protection against bloodborne pathogens.
Ordering information: Tubes with sealing caps include a retained silicone gasket in addition to the tube and screw closure. Nominal volumes correspond to rotor tube sizes; please refer to nominal volumes when ordering tubes to fit a rotor.
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty