Would you like to log in again?
Please be aware that our e-commerce platform will be undergoing scheduled maintenance from 6:00 AM to 2:00 PM ET on May 11, 2024. During this time our site will be not available. We appreciate your patience.
Nalgene® Oak Ridge Centrifuge Tubes, Polypropylene Copolymer, Thermo Scientific
Supplier: Thermo Scientific
Nalgene®
3119-0028 3139-0010 3139-0030 3139-0016 3119-0030 3119-0050 3119-0010 3139-0050
21010-523CS 947.12 USD InStock
21010-523 21010-567 21010-578 21010-524 21009-375 21009-364 21009-386 21009-397
Nalgene® Oak Ridge Centrifuge Tubes, Polypropylene Copolymer, Thermo Scientific
Tubes Centrifuge Tubes
Contact-clear, leakproof polypropylene copolymer tubes allow easy examination of contents.
  • Repeatedly autoclavable
  • Can be used upto 50000 ×g in refrigerated centrifuges
Greater chemical resistance than polycarbonate tubes. Before autoclaving, place closure on top of container without engaging threads.
Certifications: Meets OSHA Standard 29CFR, Part 1910.1030 for use as protection against bloodborne pathogens.
Ordering information: Tubes with sealing caps are supplied with polypropylene screw closure and retained silicone gasket. Tubes with screw caps are supplied with polypropylene screw closures. Nominal volumes correspond to rotor tube sizes; please refer to nominal volumes when ordering tubes to fit a rotor.
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty