Would you like to log in again?
Nalgene® Wide Mouth Centrifuge Bottle with Sealing Cap, Polypropylene Copolymer, Conical Bottom, Thermo Scientific
Supplier: Thermo Scientific
Nalgene®
3143-0175
21010-605CS 1395.84 USD InStock
21010-605
Nalgene® Wide Mouth Centrifuge Bottle with Sealing Cap, Polypropylene Copolymer, Conical Bottom, Thermo Scientific
Bottles Centrifuge Bottles
This bottle is ideal for cell harvesting and protein biochemistry applications
Note: When autoclaving, place cap loosely on bottle without engaging threads. Bottles must be used with Nalgene® adapter 21020-249. For proper performance, fill bottle to at least 80% of the total capacity.
Wide mouth with molded graduations for easy filling and decanting. Leakproof and autoclavable.
Ordering information: Supplied with polypropylene sealing cap with silicone gasket.
Order Now
ORDER
ACCESSORIES
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty