Would you like to log in again?
Nalgene® Oak Ridge Conical Centrifuge Tube Adapter; white PC, Thermo Scientific
Supplier: Thermo Scientific
DS3147-0050
21010-613CS 74.26 USD InStock
21010-613
Nalgene® Oak Ridge Conical Centrifuge Tube Adapter; white PC, Thermo Scientific
Tubes Centrifuge Tubes
Nalgene oak ridge conical tube adapter must be used with Nalgene 35 L conical oak ridge tubes.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty