Would you like to log in again?
NALGENE® Centrifuge Tube Closures, Polypropylene, Thermo Scientific
Supplier: Thermo Scientific
REVIEWS (0)
Total Ratings: 0
Avg. Ratings: out of 5
Sort all by:
Nalgene®
DS3111-0017 DS3111-0018 DS3111-0025 DS3111-0016 DS3111-0029 DS3111-0032 DS3111-0030
21010-885CS 70.56 USD InStock
21010-885 21010-943 21010-954 21010-976 21010-965 21010-911 21010-894
NALGENE® Centrifuge Tube Closures, Polypropylene, Thermo Scientific
Closures Test/Sample Tube Closures
Friction-fit polypropylene closures designed for NALGENE® round or conical centrifuge tubes
Pull tab for closure removal. Autoclavable. May not provide liquid-tight seal at high speeds. Closure 21010-965 is for use with round, nonlipped 50mL tubes.
Order Now
ORDER
WRITE A REVIEW
REVIEWS (0)
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty