Would you like to log in again?
Nalgene® Centrifuge Bottles with Caps, Polypropylene Copolymer, Thermo Scientific
Supplier: Thermo Scientific
Nalgene®
3141-0500 3141-1006 3120-1010 3120-0500 3120-0250 3120-1000 3120-9500 3141-0250
21020-040CS 508.8 USD InStock
21020-040 21020-061 21020-050 21010-636 21010-614 21020-028 89050-120 21020-152
Nalgene® Centrifuge Bottles with Caps, Polypropylene Copolymer, Thermo Scientific
Bottles Centrifuge Bottles
Wide mouth bottles are translucent and have excellent chemical resistance
Note: Before autoclaving, place cap on the container without engaging the threads. When using the 250 ml bottles (21010-614 and 21020-028) with the Sorvall® GSA 6-place rotor, use the white low-density polyethylene 250 ml adapter (21020-201). For proper performance, fill bottles to at least 80% of the total capacity (75% for 21020-050).
All caps are polypropylene. Sealing caps feature a retained silicone gasket. Both cap types are autoclavable. Bottle 89050-120 is graduated and designed for large volume gross separations, especially bacterial, yeast, and tissue isolations. It has smooth inner walls to reduce shearing, a modified polyphenylene-ether screw closure, and a 50-cycle life span. It meets USP VI requirements and is non-cytotoxic.
Order Now
ORDER
ACCESSORIES
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty