Would you like to log in again?
Nalgene® Wide Mouth Centrifuge Bottle with Screw Cap, High-Density Polyethylene, Thermo Scientific
Supplier: Thermo Scientific
Nalgene®
3121-0250
21020-072CS 498.24 USD InStock
21020-072
Nalgene® Wide Mouth Centrifuge Bottle with Screw Cap, High-Density Polyethylene, Thermo Scientific
Bottles Centrifuge Bottles
Ideal for use in refrigerated centrifuges.
Note: For proper performance, fill bottles to at least 80% of the total capacity.
Wide mouth provides easy access. Withstands temperatures to –100 °C (–148 °F). Manufactured from chemically resistant HDPE. Polypropylene screw closure.
Order Now
ORDER
ACCESSORIES
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty