Would you like to log in again?
Nalgene® Centrifuge Bottles with Caps, Polycarbonate, Thermo Scientific
Supplier: Thermo Scientific
REVIEWS (0)
Total Ratings: 0
Avg. Ratings: out of 5
Sort all by:
Nalgene®
3122-0500 3140-1006 3140-0500 3122-1000 3122-1010 3140-0250 3122-0250
21020-094CS 962.88 USD InStock
21020-094 89050-118 21020-108 21020-130 21020-163 21010-432 21010-421
Nalgene® Centrifuge Bottles with Caps, Polycarbonate, Thermo Scientific
Bottles Centrifuge Bottles
Wide mouth bottles are transparent, with excellent mechanical strength.
Screw cap for bottle 21020-108 can be replaced with sealing cap assembly 21009-901 for leakproof performance at high speeds or when hazardous materials must be contained.
Note: Before autoclaving, place cap on container without engaging threads. For proper performance, fill bottles to at least 80% of the total capacity.
All caps are polypropylene except the cap for bottle 89050-118, which is modified polyphenylene-ether. Sealing caps feature a retained silicone gasket. All bottles and caps are autoclavable.
Order Now
ORDER
ACCESSORIES
DOCUMENTATION
WRITE A REVIEW
REVIEWS (0)
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty