Would you like to log in again?
Nalgene® Top Works™ Aseptic Closure System, Silicone, for Bottles and Carboys, Thermo Scientific
Supplier: Thermo Scientific
REVIEWS (0)
Total Ratings: 0
Avg. Ratings: out of 5
Sort all by:
Nalgene® Top Works™
2135-5300 2135-5303 2132-1003 2135-8303 2135-8300 2135-3803 2135-8302 2135-5302
21020-208CS 390.22 USD InStock
21020-208 21020-206 21020-204 21020-216 21020-214 21020-212 21020-210 21020-254
Nalgene® Top Works™ Aseptic Closure System, Silicone, for Bottles and Carboys, Thermo Scientific
Closures Bottle Closures
Provides an aseptic seal for transferring high-purity fluids
Eight configurations permit precise matching of Top Works components to bottles and carboys. Polypropylene closures have center holes. They are modified to clamp a silicone stopper onto the bottle or carboy, creating a primary elastomeric seal. High-purity, platinum-cured silicone tubing ensures the fluid contact areas are crevice-free, which helps eliminate cross-contamination.
Materials are food-grade and noncytotoxic. Complies with U.S.P. Class VI requirements. Autoclavable.
For use with 125 mL to 50 L plastic bottles and carboys.
Delivery information: Solid insert units are complete with closure and insert. Two-port insert units include closure, two-port insert, and two pieces of 6.35mm (1/4") I.D. tubing. 21020-216 includes closure, three-port insert, one piece of 9.5mm (3/8") I.D. tubing, and two pieces of 6.35mm (1/4") I.D. tubing. 21020-210 includes closure, three-port insert, one piece of 3.175mm (1/8") I.D. tubing, and two pieces of 6.35mm (1/4") I.D. tubing. 21020-204 is complete with closure, three-port insert, one piece of 6.35mm (1/4") I.D. tubing, and two pieces of 3.175mm (1/8") I.D. tubing.
Caution: Note: Not for use when creating pressure or vacuum conditions with containers that are not rated for pressure or vacuum applications. Not intended for invasive use.
Order Now
ORDER
DOCUMENTATION
WRITE A REVIEW
REVIEWS (0)
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty