Would you like to log in again?
Nalgene® Centrifuge Bottle with Screw Cap, Teflon® FEP, Thermo Scientific
Supplier: Thermo Scientific
Nalgene®
3127-0250
21020-403CS 1572.48 USD InStock
21020-403
Nalgene® Centrifuge Bottle with Screw Cap, Teflon® FEP, Thermo Scientific
Bottles Centrifuge Bottles
This bottle is ideal for low speed centrifuging of aggressive chemicals or large volume lipid extractions, precipitations, and digestions.
Use Nalgene® sealing cap assembly 21009-901 to run at maximum speed.
Note: Rated for temperatures from –100 to +150 °C (–148 to +302 °F). Autoclavable. Leakproof at low speed. When autoclaving, place cap on container without engaging threads. In a refrigerated centrifuge, bottle must be run with 100% full volume and tightly capped.
The low adsorption composition is suitable for handling biomolecules, such as proteins and nucleic acids. Bottle is Teflon® FEP. Resists most acids, bases, and organic solvents. Screw cap is manufactured from Tefzel® ETFE and is leakproof.
Order Now
ORDER
ACCESSORIES
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty