Would you like to log in again?
PYREX® Heavy Duty Round Bottom Centrifuge Tubes, Corning
Supplier: Corning
PYREX®
8422-100 8422-50
21023-403CS 80.12 USD InStock
21023-403 89091-460
PYREX® Heavy Duty Round Bottom Centrifuge Tubes, Corning
Tubes Centrifuge Tubes
PYREX®, borosilicate glass, round bottom, with screw cap.
  • Excellent chemical resistance
  • High temperature resistance
  • Minimal thermal expansion, giving relatively high resistance to temperature changes
These 50 ml round bottom centrifuge tubes are useful for the handling of sputum specimens. Digestion, shaking, neutralizing and centrifuging can be performed in one container. They are also useful for other clinical and microbiological tests. Screw caps resist the effects of temperature and steam. The 50 ml tube has a black phenolic GPI 24-410 threaded cap with a glued-in, white rubber liner.
Order Now
ORDER
SPECIFICATIONS
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty