Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Microcentrifuge Tubes with Snap Cap, Bio Plas

Supplier: Bio Plas

REVIEWS (0)

Total Ratings: 0
Avg. Ratings: out of 5

Sort all by:

4156 4153 4003 4004 4025 4159 4151 4005 4157 4152 4154 4158
21150-332CS 696.69 USD InStock
21150-332 21150-342 21150-352 21150-330 21150-326 21150-348 21150-336 21150-346 21150-328 21150-338 21150-340 21150-350
Microcentrifuge Tubes with Snap Cap, Bio Plas
Tubes Microcentrifuge Tubes
The Snap-Cap Microcentrifuge Tube gives a firm, dependable seal, and its tough virgin-polypropylene construction withstands the rugged G-forces of high-speed centrifugation.

  • Ultra low retention
  • Snap-Caps for optimal sealing
  • RNase and DNase free and non-pyrogenic

Used for specimen collection, mixing, electrophoresis sampling/handling, enzyme assays, radioassays, immumoassays and many other applications. The Snap-Cap Microcentrifuge Tube’s smooth, conical interior lets remove entire sample with ease, and without material hangup. Fits Beckman™, Eppendorf™ and all other microcentrifuges.

The Snap-Cap is attached to each tube, and the entire unit is autoclavable at 250°F
at 15 psi for 15 minutes.

Order Now
 

ORDER

WRITE A REVIEW

REVIEWS (0)

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty