Would you like to log in again?
Chemstrip™ 10UA Urine Test Strips, Roche
Supplier: Roche
Chemstrip™
11895354160
21247-880EA 147.88 USD InStock
21247-880
Chemstrip™ 10UA Urine Test Strips, Roche
Clinical Diagnostic Supplies Clinical Reagents and Kits
Test strips that test for specific gravity, pH, leukocytes, nitrite, protein, glucose, ketones, urobilinogen, bilirubin, and hemoglobin in urine.
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty