Would you like to log in again?
Whatman™ Cellulose Chromatography Papers
Supplier: Whatman products (Cytiva)
Whatman™
3001-878 1030-024 3031-681 1030-025 3001-917 3454-651 3030-866 10382581 3017-917 3030-861 3030-6188 3017-8793 3001-640 3001-651 3030-6185 3031-915 3031-681 3001-651 3001-640 3001-652 3030-221 3004-8098 3003-917 3030-909 3030-917 3031-915 3030-662 3001-878 3017-621 3001-681 3030-6189 3001-652 3001-604 3017-8793 3030-6189 3004-8098 3030-672 3001-653 3001-861 3030-347 10382562 3003-917 3030-681 3001-653 3002-917 3001-917 3030-221 1030-025 3030-392 3004-614 3030-392 3454-651 3030-690 3001-861 3004-917 10382562 3030-662 3030-672 3001-614 3030-861 3001-964 3030-6132 3030-347 3030-335 3001-672 3020-917 3030-704 3020-917 3017-820 10382514 3001-917 10382581 3001-604 3001-614 30176715 10382461 3030-6132 3001-604 3030-700 3030-335 3004-614 3030-6461 3001-931 3030-700 3030-675 3001-672 3001-845 3030-931 3001-604 3017-915 3030-614 3017-621 3001-640 3001-861 3002-911 3001-652 3030-153 3030-704 3017-915 3030-931 3030-6185 3030-153 3001-640 3001-845 3017-820 3004-917 3030-690 3002-911 30176715 3030-928 3001-964 3031-901 3030-675 3001-681 1030-024 3001-931 3017-917 3030-866 3030-681 3002-917 3001-614 3030-917 3002-9159 3030-614 10382461 3030-6188 3030-6461 3002-9159 3031-901 10382514
76423-774PK 672.91 USD InStock
CA11013-876 CA21427-648 CA21427-366 CA21427-003 CA21427-364 CA21427-524 CA21427-127 CA76423-776 76423-774 76423-776 21427-240 CA21427-373 21427-364 CA28459-165 28461-000 21427-080 470205-538 CA11013-766 CA11013-006 470205-536 CA11013-764 470205-534 470205-532 21427-105 21427-626 CA11012-714 CA76423-774 CA21427-240 28459-165 CA21427-080 CA11013-758 89022-360 10035-428 CA10147-940 28452-064 21427-411 CA11013-572 CA28451-028 CA21427-386 CA21427-546 CA21427-149 CA10033-476 10035-430 CA21427-395 CA21427-393 21427-386 10035-436 CA28459-187 CA11013-608 10035-438 CA11013-606 10035-432 CA11013-604 10035-434 CA11013-742 CA28452-064 CA11013-862 21427-003 CA21427-411 21427-366 21427-127 21427-524 21427-648 28451-028 CA21426-907 10035-440 21427-373 10035-842 10035-688 28459-187 CA21426-623 10035-844 10035-442 10035-444 10035-686 CA89022-360 CA28461-000 CA21427-207 CA21427-604 470199-890 10035-846 10035-848 CA11013-792 CA21427-047 CA21427-444 21427-160 CA21427-450 21427-681 21426-907 21426-623 10034-870 10147-942 10147-940 21427-149 CA21427-397 CA21427-430 470199-888 21427-546 10034-872 CA21427-160 76480-298 21427-393 CA21427-681 21427-395 21427-397 21427-430 28297-452 CA10147-942 CA21427-626 CA11013-772 28458-005 CA11013-770 CA28297-452 21427-456 21427-458 CA21427-105 470205-528 76480-300 21427-580 CA11013-768 CA28458-005 21427-444 21427-047 CA21427-458 470005-370 CA21427-456 21427-604 21427-207 21427-450 CA21427-580
Whatman™ Cellulose Chromatography Papers
Filters Filter Paper Chromatography Paper
Whatman chromatography papers are widely used across the world, reflecting their purity, high quality and consistency. They are made from specially selected cotton cellulose and rigorously quality controlled to ensure uniformity within the grade.
  • Pure cellulose produced from the highest quality cotton linters, with no additives, ensures that no contamination will occur during the transfer steps
  • Manufactured and tested specifically for chromatographic techniques to ensure the wicking capability and uniformity of capillary action that is important in obtaining clean and even transfers during blotting
Grade 1 Chr: Smooth surface. Good resolution for general analytical separations.
Grade 2 Chr: Slower than 1 Chr for higher resolution applications. Smooth surface. Particularly recommended for optical or radiometric scanning.
Grade 3 Chr: A medium thickness paper. For general applications with medium/heavy solute loadings. Frequently used for separation of inorganic compounds and for electrophoresis.
Grade 3MM Chr: Though widely used as a blotting paper, 3MM Chr is also used both in electrophoresis and for general chemistry. A medium thickness paper used extensively for general chromatography and electrophoresis.
Grade 4 Chr: Fastest of the thin papers. Recommended for routine and/or repetitive chromatography when loadings are relatively low. Smooth surface. Very suitable where speed is important and very high resolution is not required.
Grade 17 Chr: Highly absorbent paper suitable for heavy loadings. Recommended for preparative paper chromatography and electrophoresis.
Grade 20 Chr: For maximum resolution, this grade is supreme giving the greatest possible separation of closely related compounds. Smooth surface. Recommended for separation of samples of unknown composition, with outstanding resolution at low loadings.
Grade 31ET Chr: Extremely fast. Flow rate is the highest of all chromatography papers in the Whatman range. Thick paper with fairly soft surface. Principal application is in electrophoresis of large molecules.
Grade 54 SFC: Thin hardened paper with high speed and fair to good resolution. Recommended for routine chromatography. High wet strength.
Grade 2668 Chr: For separation of relatively large molecules by electrophoresis.
Grade 2727 Chr: For separation of very large amounts of substance.
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty