Would you like to log in again?
Pallflex® Emfab™ Filters
Supplier: Cytiva
REVIEWS (0)
Total Ratings: 0
Avg. Ratings: out of 5
Sort all by:
Pallflex® Emfab™
7218 7234 7225 7224 7221 7218 7223 7219 7217 7221 7222 7224 7225 7222 7234 7223 7217 7219
28150-929PK 224.62 USD InStock
CA28150-929 28150-929 28150-923 CA21431-428 CA28150-921 28150-933 CA28150-931 21431-428 28150-921 28150-931 CA28150-925 CA28150-935 CA28150-923 28150-925 28150-935 CA28150-933 CA29301-726 29301-726
Pallflex® Emfab™ Filters
Filters Glass Fiber Filters
Pallflex Emfab filters are made of borosilicate glass microfibers that are reinforced with woven glass cloth and bonded with PTFE.
  • Glass fiber
  • Reinforced with cloth and PTFE
  • Low air resistance
These filters feature low air resistance which makes them ideal for use in critical aerosol sampling tests, such as diesel exhaust. They can also withstand folding for weighing and transport.
Ordering information: Both TX40HI45 and TX40HI75 grades have a higher level than the TX40HI20WW grade. The TX40HI75 grade has a higher level of binder than the TX40HI45.
Order Now
ORDER
SPECIFICATIONS
WRITE A REVIEW
REVIEWS (0)
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty