Would you like to log in again?
KIMAX® Chromatographic Columns, Kimble Chase
Supplier: DWK Life Sciences
KIMAX®
17800-22300 17800-19300 17800-19500 17800-11500 17800-22500 17800-11300
21503-123CS 205.21 USD InStock
21503-123 21503-156 21503-067 21503-065 21503-010 21503-015
KIMAX® Chromatographic Columns, Kimble Chase
Chromatography Columns
With removable stopcock of PTFE and replaceable glass tip.
Lower end of the column is tooled for extra strength and tight, leakproof fit. Glass tip is supplied.
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty