Would you like to log in again?
Brinkmann® TC-45 Temperature Controller, Metrohm
Supplier: Metrohm
Brinkmann®
BOM-LSTC45240
21506-120EA 1617.54 USD InStock
21506-120
Brinkmann® TC-45 Temperature Controller, Metrohm
Controllers Temperature Controllers
The low-cost TC-45 is designed to meet the needs of “above ambient” chromatography applications
Dip switches on the front panel allow for easy temperature setting in 5 °C increments from 30 to 65 °C. Constructed of sturdy 1/8" thick formed aluminum.
Certifications: UL and CSA approved.
Order Now
ORDER
SPECIFICATIONS
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty