Would you like to log in again?
Graphite Ferrules for Gas Chromatography, Ohio Valley
Supplier: Ohio Valley
GF-68 GF-03 GF-05-HP GF-08 GF-10-HP GF-04 GF-01 GF-05 GF-81 GF-02 GF-08-HP GF-64
21508-840PK 44.56 USD InStock
21508-840 21508-820 21508-829 21508-848 21508-827 21508-828 21508-822 21508-844 21508-834 21508-823 21508-846 21508-835
Graphite Ferrules for Gas Chromatography, Ohio Valley
Fittings and Connectors
The graphite ferrules fit “Swagelok” or “Parker” fittings
  • Thermally conditioned and ready for immediate high temperature GC use
  • Can be reused if not over tightened
Graphite ferrules are soft and compliant. They will
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty