Would you like to log in again?
Optimize PTFE Tubings
Supplier: Optimize Technologies
10-15-00220 10-15-00205 10-15-00206 10-15-00243 10-15-00214 10-15-00240 10-15-00238 10-15-00232 10-15-00227 10-15-00233 10-15-00226 10-15-00221 10-15-00245
21510-242PK 428.61 USD InStock
21510-242 21509-045 21509-042 10123-224 21510-588 21509-047 21509-039 21510-591 21510-241 77100-150 77100-152 21509-040 21509-051
Optimize PTFE Tubings
Tubing Chromatography Tubing
  • Various OD and ID options
  • Chemically inert to most solvents
  • Easy to cut
  • Translucent for easy monitoring
  • Good for general/ low pressure applications
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty