Would you like to log in again?
Upchurch Scientific® Thread-to-Thread Adapters, IDEX Health & Science
Supplier: IDEX Health & Science
Upchurch Scientific®
1583 1592 P-135 P-882 P-881 U-504 P-669-01 P-652 P-650 P-626 UH-630 U-500-01 P-671 U-659 P-627 P-621 P-770 1591 U-510 U-511
53498-550EA 29.96 USD InStock
53498-550 97055-124 21511-416 97055-108 97055-096 97055-110 53403-086 97055-134 21511-690 53500-702 53500-766 21511-180 21511-192 53498-526 13500-478 21511-168 21528-168 21511-178 13500-028 21511-692
Upchurch Scientific® Thread-to-Thread Adapters, IDEX Health & Science
Fittings and Connectors
IDEX Health & Science offers a wide assortment of Upchurch Scientific® threaded port adapters to help bring products with different thread configurations together.
  • High pressure ratings
  • Variety of adapters available
  • Choose from polymer and stainless steel versions
The pressure ratings of these adapters often exceed the pressure holding ability of the fittings and tubing used with them
Create a true zero dead volume connection between 1/16 in. capillary tubing with Upchurch Scientific® MicroTight™ Adapters from IDEX Health & Science in an inline connection with the ability to withstand up to 5,000 psi (345 bar), depending upon the adapter configuration. (When MicroTight tubing sleeves are used, the pressure rating lowers to 4,000 psi / 276 bar).
External National Pipe Thread Adapters make connections to female 1/8 in and 1/4 in National Pipe Thread (NPT) ports. Manufactured from PEEK™ polymer by IDEX Health & Science, Upchurch Scientific® NPT Adapters are durable and chemically resistant and available in a variety of female threads, plus a male threaded version that is suitable for most low pressure applications. Please Note: Wrap the threads on the NPT side of these adapters with thread seal tape (plumbers tape) to ensure a leak-free seal.
Choose from polymer and stainless steel versions.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty