Would you like to log in again?
Due to scheduled system maintenance, vwr.com will be temporarily unavailable from 6:00 am EST to 2:00 PM EST on Saturday, May 25. 
Bunsen Round Jaw Extension Clamps
Supplier: Humboldt
H-8562
21571-000EA 42.7 USD InStock
21571-000
Bunsen Round Jaw Extension Clamps
Clamps General Purpose Clamps
Heavy-gauge stamped steel
A concealed spring at the junction of both arms makes the jaws self-opening. With bolt and wing-nut; jaws are plastisol covered.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty