Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

VWR® Talon® Fixed-Position Clamps

Supplier: VWR

REVIEWS (0)

Total Ratings: 0
Avg. Ratings: out of 5

Sort all by:

VWR® Talon®
80862570 915870 80862569 915869
21573-266CS 1195.41 USD InStock
21573-266 89202-630 21573-265 89202-632
VWR® Talon® Fixed-Position Clamps
Clamps General Purpose Clamps
Used to hold apparatus near the lab frame where no adjustment is required after setup.

  • electropolished stainless steel finish or nickel-plated zinc
  • Use as a single buret holder
  • No adjustment required after setup
  • Two or three prongs

Built-in holder grips rods up to 19mm (3/4") in diameter. Fixed-position clamps have an integral holder but can be rotated after attachment to a lab frame or other apparatus. Available with two or three prongs. electropolished stainless steel finish or nickel-plated zinc construction.

Ordering information: Supplied with non-slip vinyl sleeves, and for temperatures above 100°C (212°F), fiberglass covers.

Order Now
 

ORDER

DOCUMENTATION

WRITE A REVIEW

REVIEWS (0)

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty