Would you like to log in again?
Bransonic® Tabletop Cleaner, Branson
Supplier: Branson
Bransonic®
100-951-010 100-951-011
21800-006EA 171.29 USD InStock
21800-006 47727-392
Bransonic® Tabletop Cleaner, Branson
Ultrasonic Cleaners
Compact ultrasonic cleaner can be conveniently plugged in anywhere
Features a stainless steel 15-ounce-capacity tank and a five-minute timer that shuts off automatically.
Cleaner has many applications, including cleaning instrument and clock parts, small electrical/electronic components, optical pieces, and recorder and drafting pens.
Ordering information: All cleaners are supplied with a cover and parts basket.
Order Now
ORDER
SPECIFICATIONS
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty