Would you like to log in again?
Please be aware that our e-commerce platform will be undergoing scheduled maintenance from 6:00 AM to 2:00 PM ET on May 11, 2024. During this time our site will be not available. We appreciate your patience.
VWR® Traceable® Nano™ Timer
Supplier: VWR
VWR® Traceable® Nano™
90013-40
21800-064EA 36.56 USD InStock
21800-064
VWR® Traceable® Nano™ Timer
Timers Digital Timers
At the press of a button, timer displays countdown alarm time/stopwatch time, and time of day in both 12- and 24-hour format.
  • Memory feature enables standards for repetitive tests
  • Digital display with highly visible digits
  • Two-channel memory storage
Timer counts up or down to 23 hours, 59 minutes, and 59 seconds on the two timing channels. The high-decibel alarm sounds for one minute. For repetitive tests, the two-channel memory automatically returns the display to the original programmed time. Digital display with 1 cm (³/₈") high digits can be read from 3.1 m (10') away.
Certifications: A serial numbered certificate is provided from an ISO 17025 calibration laboratory accredited by A2LA to indicate instrument traceability to standards provided by the National Institute of Standards and Technology.
Ordering information: Supplied with silver oxide battery, clip, stand, and rear-mounted magnet.
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty