Would you like to log in again?
VWR® Traceable™ Kangaroo Thermometer
Supplier: VWR
VWR® Traceable®
VC4430 VC1062 90012-51 VC2062
10171-326EA 84.73 USD InStock
10171-326 21800-068 10171-606 10171-490
VWR® Traceable™ Kangaroo Thermometer
Thermometers Digital Thermometers
Easy-to-use, portable unit automatically captures and recalls minimum and maximum temperature readings over any period of time.
  • Easy to use
  • Portable
  • Stainless steel probe
  • Compatible for use with most chemicals
The high/low alarm can be programmed in 1° increments and sounds every minute until the temperature returns to non-alarm conditions. The hold button freezes the display to record a current reading. Unit features large digital display with 1.9 cm (3/4") digits that are readable from 6m (20') away. Unit includes stainless steel probe with 106.7 cm (42") cable that can be stored in rear reel-compartment pouch. The probe can be used in most chemicals.
Certifications: A serial numbered certificate is provided from an ISO 17025 calibration laboratory accredited by A2LA to indicate instrument traceability to standards provided by the National Institute of Standards and Technology.
Ordering information: Supplied with silver-oxide battery that runs continuously for one year.
Order Now
ORDER
SPECIFICATIONS
SERVICES
Services: VWRCATALYST™ provides convenient and efficient re-calibration services for your digital thermometers with an industry leading 5-day average turn-around time from receipt of your instrument. All the service will be performed by an ISO17025 certified lab. Recalibration, certification and shipping fees are all included in the catalog number cost. Simply order the stated catalog number based on how many calibration points are desired. You will receive a protective box to ship your instrument to Avantor's certified calibration laboratory along with a work order form and a prepaid return shipping label. Upon receipt at the laboratory your instrument will be inspected for any defects and the performance of your instrument will be compared to certified standards. After re-calibration, your calibration report and instrument will be carefully repacked then returned with standard ground shipping. We also provide expedited service options available upon request.
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty