Would you like to log in again?
Bench and Daub Safety Can, Eagle Manufacturing
Supplier: Eagle Mfg
B606 B601 B604 B608 B606NL
21815-126EA 95.74 USD InStock
21815-126 21815-228 21815-206 21815-160 21815-181
Bench and Daub Safety Can, Eagle Manufacturing
Safety Cans
For safer cleaning of small parts or swabbing larger ones
Seamless 24-gauge terne steel construction. Dasher screen acts as fire baffle. Components are easily removed for cleaning.
Certifications: FM approved. Meets OSHA requirements.
Ordering information: Safety can 21815-206 is supplied without lid.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty