Would you like to log in again?
CLEANER SOLUTION BUFFCMP 1GL
Supplier: BRANSON ULTRASONICS CORP.
BC Concentrated Solution^ Biodegradable, phosphate and caustic free cleaner for removing buffing compound. Blend of non-ionic surfactants, detergents, and emulsifiers. Non-viscous liquid rapidly removes tripoli, rouge, lime, diamond tripoli from buffed and polished components.
ORDER
Expand All / Collapse All
Description VWR Catalog Number Supplier No Unit Price Quantity
CLEANER SOLUTION BUFFCMP 1GL 21830-287 000-955-316 Case of 4 (1/ Itm/Bx) Retrieving
Each(1/Itm/Bx) Retrieving
Green Product
Generate Barcodes Barcode Label Format: 
Generate PDF Catalog Page
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty