Would you like to log in again?
MICRO-90® Concentrated Cleaning Solution, International Products
Supplier: International Products
MICRO-90®
M-9050-12 M-9033 M-9034 M-9051-04
89210-138CS 349.44 USD InStock
89210-138 21830-422 89210-140 21830-420
MICRO-90® Concentrated Cleaning Solution, International Products
Cleaning Agents Detergents and Surface Cleansers
A powerful, multi-purpose liquid for precision cleaning of glassware, filter membranes, laboratory equipment, electronics, metal parts, plastics, and other surfaces
  • Mildly alkaline, aqueous cleaner lifts, disperses, emulsifies, sequesters, dissolves, suspends, and decomposes soils
  • Concentrate mixes instantly with water
  • Safe to use
  • Cleaner is environmentally friendly, with no CFCs, ODCs, phosphates, or phenols
Highly-concentrated — a one liter container of cleaner makes over 12 gallons at 2% solution.
Best used in manual, immersion, and low-pressure or low-agitation systems. Ideal for use in ultrasonic cleaners.
Certifications: Cleaner is NSF-registered as an A1, general cleaner in and around food-processing areas and equipment.
Ordering information: Validation methods are available upon request.
Order Now
ORDER
SPECIFICATIONS
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty