Would you like to log in again?
Citranox® Acid Cleaner and Detergents
Supplier: Alconox
REVIEWS (0)
Total Ratings: 0
Avg. Ratings: out of 5
Sort all by:
Citranox®
1855 1801 1832 1815 1801 1815 1855
75801-042CS 293.25 USD InStock
75801-042 21835-353 CA21835-350 21835-350 21835-356 CA21835-353 CA21835-356
Citranox® Acid Cleaner and Detergents
Cleaning Agents Detergents and Surface Cleansers
CITRANOX is a phosphate free, concentrated cleaner and metal brightener for high performance removal of metal oxides, scale, salts and inorganic residues in manual and ultrasonic cleaning. Corrosion inhibited and free rinsing for reliable results and no interfering residues. Ideal for trace metal analysis.
  • Concentrated to save you money
  • Phosphate free, biodegradable and readily disposable
  • Penetrating wetting power to save you time
  • Free rinsing to give you reliable results and no interfering residues
  • Use to pass your critical cleaning validation tests for lab accreditation and plant inspection approval
  • Dilute 2:100
For removing scale, metal oxides, metal complexes, trace inorganics, milkstone, pasta and starch, soil, grit, buffing compound, grime, grease, fats, oils, particulates, deposits, chemical and solvents.
They can be used to clean airy equipment, laboratory ware, sampling apparatus, lab stills, laser chillers, clean rooms, optical parts, electronic components, pharmaceutical apparatus, cosmetics manufacturing equipment, restaurant equipment, rethermolators, steam tables, metal forgings, stampings and castings, industrial parts, pipes, tanks, boilers and reactors.
Cleaning method: Soak, brush, sponge, cloth, ultrasonic cleaner, flow through clean-inplace.
Corrosion inhibited formulation recommended for glass, metal, stainless steel, porcelain, ceramic, plastic, rubber and fiberglass. Can be used on soft metals such as copper, bronze, aluminum, and zinc if rinsed promptly.
Note: Corrosion testing is advisable.
Certifications: Authorized by USDA for use in federally inspected meat and poultry plants. FDA certified.
Caution: Will foam—not for spray or machine use.
Order Now
ORDER
DOCUMENTATION
WRITE A REVIEW
REVIEWS (0)
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty