Would you like to log in again?
Please be aware that our e-commerce platform will be undergoing scheduled maintenance from 6:00 AM to 2:00 PM ET on May 11, 2024. During this time our site will be not available. We appreciate your patience.
Clorox® Healthcare® Bleach Germicidal Cleaner
Supplier: Essendant
Clorox®
CLO 68967 CLO68978 CLO68970
21850-206CS 378.15 USD InStock
21850-206 21850-204 21850-210
Clorox® Healthcare® Bleach Germicidal Cleaner
Cleaning Agents Disinfectants
Healthcare professionals trust Clorox® Healthcare® bleach germicidal cleaner to help them overcome infection control challenges in a constantly changing healthcare environment.
  • No pre-cleaning required
  • Designed for easy refilling to meet needs of heavy users
  • EPA registered to kill a total of 50 micro-organisms
  • Proven efficacy against outbreak-causing viruses Influenza A and B viruses, Measles virus and Norovirus
  • Kill HAI-causing pathogens including Clostridioides difficile spores, MRSA and VRE in 3 minutes or less
  • Broad Spectrum disinfection efficacy against over 50 pathogens
  • 3 minutes contact time to kill Clostridioides difficile spores - tested in the presence of 3-part organic soil load following the latest EPA testing recommendations (Nov. 2016)
  • Feature an anti-corrosion agent to ensure compatibility with healthcare surfaces
  • Tested for surface safety on common healthcare and office surfaces and equipment
Clorox® Healthcare® Bleach Germicidal Cleaner is a ready to use, 1 step cleaner- disinfectant containing a 1:10 bleach dilution to meet CDC and OSHA guidelines for disinfecting surfaces in healthcare settings. This cleaner disinfectant is EPA registered to kill a total of 50 micro-organisms, including HAI causing bacteria like MRSA and VRE, outbreak causing viruses such as Influenza A and B viruses, Measles virus and Norovirus and fungi in 1 minute.
This disinfectant kill 99.9999% of Clostridioides difficile spores in 3 minutes, according to the current EPA recommended testing standard (June 2014). The ready-to-use disinfectant eliminates compliance risks and labor costs associated with dilatable disinfectants. Designed for easy refilling to meet needs of heavy users. This bleach solution features an anti-corrosion agent and is formulated for broad surface compatibility with healthcare surfaces including stainless steel, glass, plastic, porcelain, glazed ceramic, laminated surfaces and vinyl. Intended for use in healthcare settings. For use against bacteria, fungus, germs, grease, grime, mildew, mold, organic matter, virus, water-based soil. Works on ceramics, cement/concrete, enamel, fiberglass, glass, grout, laminates, metal, plastic, sealed/engineered stone, vinyl.
Certifications: Meets CDC and OSHA recommendations.
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty