Would you like to log in again?
Dust-Off® Pressurized Dusters, Falcon Safety Products®
Supplier: Falcon Safety
Dust-Off®
DPNRCN DPSXL DPNXL DPVC DPSJB DPNR FGSRCN FGSRX DPN DPNCN
21899-103CS 1552.63 USD InStock
21899-103 21899-090 21899-080 CA21899-090 21899-092 CA21899-092 21899-093 21899-094 CA21899-093 21899-130
Dust-Off® Pressurized Dusters, Falcon Safety Products®
Sprays
Provide a clean, moisture-free cleaning blast. Contain no CFCs, HCFCs, propane, or butane. Dusters are 100% ozone-safe. Disposable dusters are a larger, value size and are ideal for use with workstations, labs, repair benches, and more.
  • Disposable dusters are ideal for workstations, labs, repair benches, and more
  • Dust-Off XL is for use where exposure to potential ignition sources is a concern
  • Dust-Off Plus XL is nonflammable
  • 360° Vector Valve® allows the spray to be directed in any direction while the can remains upright
Dust-Off® XL duster 21899-080 is designed for general dusting away from laboratories where exposure to Bunsen burners and other potential ignition sources is a concern. It is flammable under certain circumstances and is formulated for safe and effective cleaning of sensitive areas, such as chemical labs.
Dust-Off Plus® XL duster 21899-094 is nonflammable.
The reusable Dust-Off® Plus duster 21899-090 includes a nozzle and a replaceable canister and is specially designed for applications where flammability is a concern. The 360° Vector Valve® allows the spray to be directed in any direction while the can remains upright.
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty